Skip to Content
Merck
All Photos(1)

Key Documents

60987

Sigma-Aldrich

Spearmint oil

natural, from Mentha spicata L.

Synonym(s):

Mentha spicata

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

biological source

Mentha spicata L.

quality

natural

refractive index

n20/D 1.489 (lit.)
n20/D 1.50

bp

228 °C (lit.)

density

0.950 g/cm3

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Spearmint oil is extracted from aerial parts of Mentha spicata L. Spearmint oil has antimicrobial property. This essential oil is used in toothpaste as a flavoring ingredient. In addition, spearmint oil is also used in chewing gum and medicines.

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allergic contact cheilitis from spearmint oil.
Skrebova N, et al.
Contact Dermatitis, 39(1), 35-36 (1998)
Chemical composition, and antioxidant and antimicrobial activities of essential oil of spearmint (Mentha spicata L.) from Pakistan.
Hussain A I, et al.
J. Essent. Oil Res., 22(1), 78-84 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service