Skip to Content
Merck
All Photos(1)

Key Documents

449105

Sigma-Aldrich

Tris(hydroxymethyl-d3)amino-d2-methane

98 atom % D, 98% (CP)

Synonym(s):

TRIS-d11

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(DOCD2)3CND2
CAS Number:
Molecular Weight:
132.20
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Assay

98% (CP)

technique(s)

NMR: suitable
bio NMR: suitable

bp

219-220 °C/10 mmHg (lit.)

mp

162-165 °C (lit.)

mass shift

M+11

SMILES string

[2H]OC([2H])([2H])C(N([2H])[2H])(C([2H])([2H])O[2H])C([2H])([2H])O[2H]

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2/i1D2,2D2,3D2,6D,7D,8D/hD2

InChI key

LENZDBCJOHFCAS-RNVFPDRBSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service