Skip to Content
Merck
All Photos(2)

Key Documents

294063

Sigma-Aldrich

Dimethyl(methylthio)sulfonium tetrafluoroborate

97%

Synonym(s):

DMTSF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SS(CH3)2BF4
CAS Number:
Molecular Weight:
196.04
Beilstein:
3572194
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

89-92 °C (lit.)

storage temp.

−20°C

SMILES string

CS[S+](C)C.F[B-](F)(F)F

InChI

1S/C3H9S2.BF4/c1-4-5(2)3;2-1(3,4)5/h1-3H3;/q+1;-1

InChI key

LUOOXKXNWLQGLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service