Skip to Content
Merck
All Photos(3)

Key Documents

284688

Sigma-Aldrich

Decanoic anhydride

96%

Synonym(s):

Capric anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)8CO]2O
CAS Number:
Molecular Weight:
326.51
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

refractive index

n20/D 1.44 (lit.)

bp

125-140 °C/0.05 mmHg (lit.)

mp

24-25 °C (lit.)

solubility

chloroform: soluble 100 mg/mL, clear, colorless to light yellow

density

0.886 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCC(=O)OC(=O)CCCCCCCCC

InChI

1S/C20H38O3/c1-3-5-7-9-11-13-15-17-19(21)23-20(22)18-16-14-12-10-8-6-4-2/h3-18H2,1-2H3

InChI key

HTWWKYKIBSHDPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Decanoic anhydride was used in the synthesis of 2-deoxy-2-p-methoxybenzylimideneaminio-1,3,4,6-tetra-O-decanoyl-D-glucopyranose.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Paolo Neviani et al.
Journal of nucleic acids, 2010 (2010-08-28)
Here we investigate the supramolecular polymerizations of two lipophilic guanosine derivatives in chloroform by light scattering technique and TEM experiments. The obtained data reveal the presence of several levels of organization due to the hierarchical self-assembly of the guanosine units
A novel extractant based on D-glucosamine for the extraction of molybdenum and tungsten.
Solvent Extraction Research and Development, Japan, 16, 47-56 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service