Skip to Content
Merck
All Photos(1)

Key Documents

181889

Sigma-Aldrich

Poly(ethyl acrylate) solution

average Mw ~95,000 by GPC, in toluene

Synonym(s):

Acrylic acid ethyl ester polymer, Ethyl acrylate homopolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

1.2 (vs air)

Quality Level

form

liquid

autoignition temp.

997 °F

mol wt

average Mw ~95,000 by GPC

composition

solids, 18-22 wt. %

concentration

in toluene

weight

Unit weight does not include weight of solvent.

refractive index

n20/D 1.469

transition temp

Tg −23 °C

density

1.21 g/mL at 25 °C

SMILES string

CCOC(=O)C=C

InChI

1S/C5H8O2/c1-3-5(6)7-4-2/h3H,1,4H2,2H3

InChI key

JIGUQPWFLRLWPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Biocompatible PEA based copolymer membranes may find several applications such as keratoprosthesis.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4.0 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Biointegration of corneal macroporous membranes based on poly (ethyl acrylate) copolymers in an experimental animal model.
Alio del Barrio J L, et al.
Journal of Biomedical Materials Research Part A, 103(3), 1106-1118 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service