Merck
  • X-Neu5Ac: a novel substrate for chromogenic assay of neuraminidase activity in bacterial expression systems.
X-Neu5Ac: a novel substrate for chromogenic assay of neuraminidase activity in bacterial expression systems.
Bioorganic & medicinal chemistry (1993-08-01)
I Fujii, Y Iwabuchi, T Teshima, T Shiba, M Kikuchi
ABSTRACT
A chromogenic substrate 1, 5-bromo-4-chloroindol-3-yl 5-acetamido-3,5-dideoxy-alpha-D-glycero-D-galacto-2-nonulopyranosidon ic acid (X-Neu5Ac), has been synthesized to facilitate the screening of bacterial colonies or plaques for the detection of either natural or mutant neuraminidase activity. Substrate 1 was hydrolyzed by neuraminidase isolated from Clostridium perfringens to release a halogenated indol-3-ol 2 that undergoes rapid aerobic oxidation to form the dark blue pigment, 5,5'-dibromo-4,-4'-dichloroindigo 3. Preliminary kinetic studies indicate that this compound is a good substrate (Km 0.89 x 10(-3) M) for neuraminidase and is quite stable under identical conditions in the absence of enzyme. These results suggest that X-Neu5Ac 1 can be useful to screen for bacterially-encoded enzyme production directly on agar plates.
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
Sigma-Aldrich
5-Bromo-4-chloro-3-indolyl α-D-N-acetylneuraminic acid sodium salt, ≥90%
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)