Merck
  • A novel method for the production of transgenic cloned pigs: electroporation-mediated gene transfer to non-cultured cells and subsequent selection with puromycin.
A novel method for the production of transgenic cloned pigs: electroporation-mediated gene transfer to non-cultured cells and subsequent selection with puromycin.
Biology of reproduction (2004-09-24)
Satoshi Watanabe, Masaki Iwamoto, Shun-ichi Suzuki, Daiichiro Fuchimoto, Daisuke Honma, Takashi Nagai, Michiko Hashimoto, Satoko Yazaki, Masahiro Sato, Akira Onishi
ABSTRACT
Puromycin N-acetyl transferase gene (pac), of which the gene product catalyzes antibiotic puromycin (an effective inhibitor of protein synthesis), has been widely used as a dominant selection marker in embryonic stem (ES) cell-mediated transgenesis. The present study is the first to report on the usefulness of puromycin for production of enhanced green fluorescent protein (EGFP) transgenic piglets after somatic cell cloning and embryo transfer. Somatic cells isolated from porcine fetuses at 73 days of gestation were immediately electroporated with a transgene (pCAG-EGFPac) carrying both EGFP cDNA and pac. This procedure aims to avoid aging effects thought to be generated during cell culture. The recombinant cells were selected with puromycin at a low concentration (2 microg/ml), cultured for 7 days, and then screened for EGFP expression before somatic cell cloning. The manipulated embryos were transplanted into the oviducts of 14 foster mother sows. Four of the foster sows became pregnant and nine piglets were delivered. Of the nine piglets, eight died shortly after birth and one grew healthy after weaning. Results indicate that puromycin can be used for the selection of recombinant cells from noncultured cells, and moreover, may confer the production of genetically engineered newborns via nuclear transfer techniques in pigs.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture