Merck
  • Design rules for light-emitting electrochemical cells delivering bright luminance at 27.5 percent external quantum efficiency.
Design rules for light-emitting electrochemical cells delivering bright luminance at 27.5 percent external quantum efficiency.
Nature communications (2017-11-01)
Shi Tang, Andreas Sandström, Petter Lundberg, Thomas Lanz, Christian Larsen, Stephan van Reenen, Martijn Kemerink, Ludvig Edman
ABSTRACT
The light-emitting electrochemical cell promises cost-efficient, large-area emissive applications, as its characteristic in-situ doping enables use of air-stabile electrodes and a solution-processed single-layer active material. However, mutual exclusion of high efficiency and high brightness has proven a seemingly fundamental problem. Here we present a generic approach that overcomes this critical issue, and report on devices equipped with air-stabile electrodes and outcoupling structure that deliver a record-high efficiency of 99.2 cd A
Sigma-Aldrich
Tetrahexylammonium tetrafluoroborate, ≥97.0%