Merck
  • The BEACH protein LRBA is required for hair bundle maintenance in cochlear hair cells and for hearing.
The BEACH protein LRBA is required for hair bundle maintenance in cochlear hair cells and for hearing.
EMBO reports (2017-09-13)
Christian Vogl, Tanvi Butola, Natja Haag, Torben J Hausrat, Michael G Leitner, Michel Moutschen, Philippe P Lefèbvre, Carsten Speckmann, Lillian Garrett, Lore Becker, Helmut Fuchs, Martin Hrabe de Angelis, Sandor Nietzsche, Michael M Kessels, Dominik Oliver, Matthias Kneussel, Manfred W Kilimann, Nicola Strenzke
ABSTRACT
Lipopolysaccharide-responsive beige-like anchor protein (LRBA) belongs to the enigmatic class of BEACH domain-containing proteins, which have been attributed various cellular functions, typically involving intracellular protein and membrane transport processes. Here, we show that LRBA deficiency in mice leads to progressive sensorineural hearing loss. In LRBA knockout mice, inner and outer hair cell stereociliary bundles initially develop normally, but then partially degenerate during the second postnatal week. LRBA deficiency is associated with a reduced abundance of radixin and Nherf2, two adaptor proteins, which are important for the mechanical stability of the basal taper region of stereocilia. Our data suggest that due to the loss of structural integrity of the central parts of the hair bundle, the hair cell receptor potential is reduced, resulting in a loss of cochlear sensitivity and functional loss of the fraction of spiral ganglion neurons with low spontaneous firing rates. Clinical data obtained from two human patients with protein-truncating nonsense or frameshift mutations suggest that LRBA deficiency may likewise cause syndromic sensorineural hearing impairment in humans, albeit less severe than in our mouse model.
Sigma-Aldrich
Phalloidin-Atto 488, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Radixin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-SLC9A3R2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Glutamate Receptor 2 & 3 Antibody, Chemicon®, from rabbit