Merck
  • PP2A Controls Genome Integrity by Integrating Nutrient-Sensing and Metabolic Pathways with the DNA Damage Response.
PP2A Controls Genome Integrity by Integrating Nutrient-Sensing and Metabolic Pathways with the DNA Damage Response.
Molecular cell (2017-06-27)
Elisa Ferrari, Christopher Bruhn, Marta Peretti, Corinne Cassani, Walter Vincenzo Carotenuto, Mohamed Elgendy, Ghadeer Shubassi, Chiara Lucca, Rodrigo Bermejo, Mario Varasi, Saverio Minucci, Maria Pia Longhese, Marco Foiani
ABSTRACT
Mec1
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Sigma-Aldrich
Dihydroceramide C2, ≥97% (TLC), solid
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Myriocin from Mycelia sterilia, ≥98% (HPLC), powder
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Cerulenin, ≥98% (HPLC), from Cephalosporium caerulens
Sigma-Aldrich
Cycloleucine, 97%