Merck
Degradation and disposal of some enzyme inhibitors. Scientific note.
Applied biochemistry and biotechnology (1994-08-01)
G Lunn, E B Sansone
ABSTRACT
Five enzyme inhibitors (phenylmethylsulfonyl fluoride, 4-amidinophenylmethanesulfonyl fluoride, 4-(2-aminoethyl)benzenesulfonyl fluoride, N alpha-p-tosyl-L-lysine chloromethyl ketone, and N-tosyl-L-phenylalanine chloromethyl ketone) in buffer, DMSO, or stock solutions were completely degraded by adding 1M NaOH and the final reaction mixtures were not mutagenic. The stability of these compounds decreased as the pH increased.
Sigma-Aldrich
AEBSF Ready Made Solution
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)