- Effect of Quaternary Ammonium Salts with Fluorine Atoms on Selected Weed Species.
Effect of Quaternary Ammonium Salts with Fluorine Atoms on Selected Weed Species.
Bulletin of environmental contamination and toxicology (2017-02-16)
Robert Biczak, Barbara Pawłowska, Maciej Płatkowski, Michał Stręk, Arkadiusz Telesiński
PMID28197704
ABSTRACT
This study investigated the effects of four structurally different quaternary ammonium salts (QASs), i.e., tetrabutylammonium tetrafluoroborate [TBA][BF