Merck
  • Integrins αvβ5 and αvβ3 promote latent TGF-β1 activation by human cardiac fibroblast contraction.
Integrins αvβ5 and αvβ3 promote latent TGF-β1 activation by human cardiac fibroblast contraction.
Cardiovascular research (2014-03-19)
Vincent Sarrazy, Anne Koehler, Melissa L Chow, Elena Zimina, Chen X Li, Hideyuki Kato, Christopher A Caldarone, Boris Hinz
ABSTRACT
Pathological tissue remodelling by myofibroblast contraction is a hallmark of cardiac fibrosis. Myofibroblasts differentiate from cardiac fibroblasts under the action of transforming growth factor-β1 (TGF-β1), which is secreted into the extracellular matrix as a large latent complex. Integrin-mediated traction forces activate TGF-β1 by inducing a conformational change in the latent complex. The mesenchymal integrins αvβ5 and αvβ3 are expressed in the heart, but their role in the activation of TGF-β1 remains elusive. Here, we test whether targeting αvβ5 and αvβ3 integrins reduces latent TGF-β1 activation by cardiac fibroblasts with the goal to prevent the formation of α-smooth muscle actin (α-SMA)-expressing cardiac myofibroblasts and their contribution to fibrosis. Using a porcine model of induced right ventricular fibrosis and pro-fibrotic culture conditions, we show that integrins αvβ5 and αvβ3 are up-regulated in myofibroblast-enriched fibrotic lesions and differentiated cultured human cardiac myofibroblasts. Both integrins autonomously contribute to latent TGF-β1 activation and myofibroblast differentiation, as demonstrated by function-blocking peptides and antibodies. Acute blocking of both integrins leads to significantly reduced TGF-β1 activation by cardiac fibroblast contraction and loss of α-SMA expression, which is restored by adding active TGF-β1. Manipulating integrin protein levels in overexpression and shRNA experiments reveals that both integrins can compensate for each other with respect to TGF-β1 activation and induction of α-SMA expression. Integrins αvβ5 and αvβ3 both control myofibroblast differentiation by activating latent TGF-β1. Pharmacological targeting of mesenchymal integrins is a possible strategy to selectively block TGF-β1 activation by cardiac myofibroblasts and progression of fibrosis in the heart.
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Integrin β5 Antibody, CT/cytosolic, serum, Chemicon®
Sigma-Aldrich
Anti-Integrin β3 Antibody, clone BB10, clone BB10, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Actin (α-Sarcomeric) antibody produced in mouse, clone 5C5, ascites fluid