Merck
  • The Arabidopsis alkaline ceramidase TOD1 is a key turgor pressure regulator in plant cells.
The Arabidopsis alkaline ceramidase TOD1 is a key turgor pressure regulator in plant cells.
Nature communications (2015-01-17)
Li-Yu Chen, Dong-Qiao Shi, Wen-Juan Zhang, Zuo-Shun Tang, Jie Liu, Wei-Cai Yang
ABSTRACT
Turgor pressure plays pivotal roles in the growth and movement of walled cells that make up plants and fungi. However, the molecular mechanisms regulating turgor pressure and the coordination between turgor pressure and cell wall remodelling for cell growth remain poorly understood. Here, we report the characterization of Arabidopsis TurgOr regulation Defect 1 (TOD1), which is preferentially expressed in pollen tubes and silique guard cells. We demonstrate that TOD1 is a Golgi-localized alkaline ceramidase. tod1 mutant pollen tubes have higher turgor than wild type and show growth retardation both in pistils and in agarose medium. In addition, tod1 guard cells are insensitive to abscisic acid (ABA)-induced stomatal closure, whereas sphingosine-1-phosphate, a putative downstream component of ABA signalling and product of alkaline ceramidases, promotes closure in both wild type and tod1. Our data suggest that TOD1 acts in turgor pressure regulation in both guard cells and pollen tubes.
Sigma-Aldrich
Sphingosine 1-phosphate, ≥95%, powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Avanti
C12-NBD Phytosphingosine, N-[12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl]-phytosphingosine, powder
Avanti
C12-NBD Ceramide, Avanti Research - A Croda Brand 810211P, powder
Avanti
C12-NBD Ceramide, Avanti Research - A Croda Brand 810211C
Avanti
Sphingosine (d18:1), Avanti Research - A Croda Brand