Merck
  • Quantitative proteomic analysis of human embryonic stem cell differentiation by 8-plex iTRAQ labelling.
Quantitative proteomic analysis of human embryonic stem cell differentiation by 8-plex iTRAQ labelling.
PloS one (2012-06-23)
Mahdieh Jadaliha, Hyoung-Joo Lee, Mohammad Pakzad, Ali Fathi, Seul-Ki Jeong, Sang-Yun Cho, Hossein Baharvand, Young-Ki Paik, Ghasem Hosseini Salekdeh
ABSTRACT
Analysis of gene expression to define molecular mechanisms and pathways involved in human embryonic stem cells (hESCs) proliferation and differentiations has allowed for further deciphering of the self-renewal and pluripotency characteristics of hESC. Proteins associated with hESCs were discovered through isobaric tags for relative and absolute quantification (iTRAQ). Undifferentiated hESCs and hESCs in different stages of spontaneous differentiation by embryoid body (EB) formation were analyzed. Using the iTRAQ approach, we identified 156 differentially expressed proteins involved in cell proliferation, apoptosis, transcription, translation, mRNA processing, and protein synthesis. Proteins involved in nucleic acid binding, protein synthesis, and integrin signaling were downregulated during differentiation, whereas cytoskeleton proteins were upregulated. The present findings added insight to our understanding of the mechanisms involved in hESC proliferation and differentiation.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Rat IgG (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer