Merck
  • Self-diffusion and interactions in mixtures of imidazolium bis(mandelato)borate ionic liquids with polyethylene glycol: (1) H NMR study.
Self-diffusion and interactions in mixtures of imidazolium bis(mandelato)borate ionic liquids with polyethylene glycol: (1) H NMR study.
Magnetic resonance in chemistry : MRC (2015-04-10)
Andrei Filippov, Nail Azancheev, Mamoun Taher, Faiz Ullah Shah, Pauline Rabét, Sergei Glavatskih, Oleg N Antzutkin
ABSTRACT
We used (1) H nuclear magnetic resonance pulsed-field gradient to study the self-diffusion of polyethylene glycol (PEG) and ions in a mixture of PEG and imidazolium bis(mandelato)borate ionic liquids (ILs) at IL concentrations from 0 to 10 wt% and temperatures from 295 to 370 K. PEG behaves as a solvent for these ILs, allowing observation of separate lines in (1) H NMR spectra assigned to the cation and anion as well as to PEG. The diffusion coefficients of PEG, as well as the imidazolium cation and bis(mandelato)borate (BMB) anion, differ under all experimental conditions tested. This demonstrates that the IL in the mixture is present in at least a partially dissociated state, while the lifetimes of the associated states of the ions and ions with PEG are less than ~30 ms. Generally, increasing the concentration of the IL leads to a decrease in the diffusion coefficients of PEG and both ions. The diffusion coefficient of the anion is less than that of the cation; the molecular mass dependence of diffusion of ions can be described by the Stokes-Einstein model. NMR chemical shift alteration analysis showed that the presence of PEG changes mainly the chemical shifts of protons belonging to imidazole ring of the cation, while chemical shifts of protons of anions and PEG remain unchanged. This demonstrated that the imidazolium cation interacts mainly with PEG, which most probably occurs through the oxygen of PEG and the imidazole ring. The BMB anion does not strongly interact with PEG, but it may be indirectly affected by PEG through interaction with the cation, which directly interacts with PEG.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Mandelic acid, 99%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
1-Methylimidazole, ReagentPlus®, 99%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
1-Methylimidazole, ≥99%, purified by redistillation
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
1-Chlorododecane, ≥97.0% (GC)
Sigma-Aldrich
1-Bromodecane, 98%
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)