Merck
  • Design of a Heterogeneous Catalyst Based on Cellulose Nanocrystals for Cyclopropanation: Synthesis and Solid-State NMR Characterization.
Design of a Heterogeneous Catalyst Based on Cellulose Nanocrystals for Cyclopropanation: Synthesis and Solid-State NMR Characterization.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-07-17)
Jiquan Liu, Andreas Plog, Pedro Groszewicz, Li Zhao, Yeping Xu, Hergen Breitzke, Annegret Stark, Rudolf Hoffmann, Torsten Gutmann, Kai Zhang, Gerd Buntkowsky
ABSTRACT
Heterogeneous dirhodium(II) catalysts based on environmentally benign and biocompatible cellulose nanocrystals (CNC-Rh2) as support material were obtained by ligand exchange between carboxyl groups on the CNC surface and Rh2(OOCCF3)4, as was confirmed by solid-state (19)F and (13)C NMR spectroscopy. On average, two CF3COO(-) groups are replaced during ligand exchange, which is consistent with quantitative analysis by a combination of (19)F NMR spectroscopy and thermogravimetry. CNC-Rh2 catalysts performed well in a model cyclopropanation reaction, in spite of the low dirhodium(II) content on the CNC surface (0.23 mmol g(-1)). The immobilization through covalent bonding combined with the separate locations of binding positions and active sites of CNC-Rh2 guarantees a high stability against leaching and allows the recovery and reuse of the catalyst during the cyclopropanation reaction.
Sigma-Aldrich
Allyl methyl sulfone, 96%
Sigma-Aldrich
Ethyl diazoacetate, contains ≥13 wt. % dichloromethane
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
TEMPO, 98%
Sigma-Aldrich
TEMPO, purified by sublimation, 99%
Sigma-Aldrich
Ethyl diazoacetate solution, 15% in toluene