Merck
Current Approach in the Diagnosis and Management of Uveitic Glaucoma.
BioMed research international (2015-11-12)
Francisco J Muñoz-Negrete, Javier Moreno-Montañés, Paula Hernández-Martínez, Gema Rebolleda
ABSTRACT
Uveitic glaucoma (UG) typically is associated with very high intraocular pressure (IOP) and more intense optic nerve damage than other glaucoma types. This secondary glaucoma requires an early diagnosis and adequate management of both uveitis and glaucoma. It is mandatory to identify the mechanisms of IOP elevation that in many eyes have multiple combined mechanisms. Management of these patients commonly requires an interdisciplinary approach that includes a glaucoma specialist and rheumatologist to control the inflammation and IOP. Glaucoma surgery is required early in these patients due to the high IOP usually present and is less successful than in primary open-angle glaucoma. Recurrent uveitic episodes, multiple mechanism, and the complications associated with uveitis make surgical management of UG challenging. In this review, the management and treatment of UG are updated to clarify the pathogenesis and prevent optic nerve damage.
Product Description
Neodymium, rod, 50mm, diameter 12.5mm, cast, 99%
Neodymium, rod, 50mm, diameter 6.35mm, cast, 99%
Neodymium, rod, 100mm, diameter 12.5mm, cast, 99%
Neodymium, rod, 100mm, diameter 6.35mm, cast, 99%
Sigma-Aldrich
Neodymium, powder, −40 mesh, ≥99% trace rare earth metals basis