Merck
  • Measurement of shrinkage and cracking in lyophilized amorphous cakes. Part I: final-product assessment.
Measurement of shrinkage and cracking in lyophilized amorphous cakes. Part I: final-product assessment.
Journal of pharmaceutical sciences (2014-11-26)
Sabine Ullrich, Stefan Seyferth, Geoffrey Lee
ABSTRACT
An experimental technique is presented to determine independently shrinkage and cracking in lyophilized amorphous cakes based on photographic imaging of their top surface. An inverse correlation between cake shrinkage and cracking during freeze-drying is seen. Shrinkage relaxes the drying tension and gives little cracking, whereas if shrinkage is restrained then more cracking occurs. A lower shrinkage and greater cracking with higher disaccharide concentration correlates with change in cake hardness and brittleness. Adhesion of the cake to the inside vial wall could not be identified as a determining factor for shrinkage. Shrinkage is non-uniform across the cake's surface and is manifested largely in the peripheral region. A correlation between shrinkage and w(g)' for different disaccharides suggests that drying tension develops as non-frozen water is lost from the porous solid after sublimation of the ice phase has exposed the solid/gas interface.
Product Description
Trehalose dihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(+)-Trehalose dihydrate, ≥99% (HPLC), from starch
Millipore
D-(+)-Trehalose dihydrate, suitable for microbiology, ≥99.0%, composed of two α-glucose units that is used as a protectant, stabilizer and to support proper folding
Sigma-Aldrich
D-(+)-Trehalose dihydrate, ≥98.5% (HPLC), from Saccharomyces cerevisiae
Sigma-Aldrich
D-(+)-Trehalose dihydrate, ≥99% (HPLC), from Saccharomyces cerevisiae
Sigma-Aldrich
D-(+)-Trehalose dihydrate, from Saccharomyces cerevisiae, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Maltose solution, for molecular biology, BioReagent, ~20% in H2O
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard