Merck
  • In vitro differentiation of human liver-derived stem cells with mesenchymal characteristics into immature hepatocyte-like cells.
In vitro differentiation of human liver-derived stem cells with mesenchymal characteristics into immature hepatocyte-like cells.
Transplantation proceedings (2014-06-18)
J-H Lee, H-J Park, I K Jang, H-E Kim, D-H Lee, J-K Park, S-K Lee, H-H Yoon
ABSTRACT
Liver transplantation is severely limited by donor shortage although it is the only effective treatment for end-stage liver disease. So the best alternative is hepatocyte transplantation. For obtaining human hepatocytes, some stem cells originating from extrahepatic or intraheptic tissues have been isolated and characterized. Previously we have reported that human liver-derived stem cells (HLSCs) could be isolated and expanded from donated livers unsuitable for transplantation; they expressed some markers of mesenchymal stem cells but neither hematopoietic nor oval cells. In this study, we isolated and expanded HLSCs with mesenchymal characteristics from another adult human liver. They showed mesenchymal morphology and grew well under serum condition similar to our previous reports. Also, they expressed some markers of mesenchymal stem cells, such as CD44, CD73, CD90, and CD105, through fluorescence-activated cell sorting analysis. When HLSCs were sequentially exposed to fibroblast growth factor-1 (FGF-1), FGF-4, and hepatocyte growth factor (HGF) followed by FGF-4, HGF, oncostatin M, and dexamethasone, they became round or polygonal, and expressed some hepatic markers such as albumin and α1-antitrypsin in the gene or protein level. Also, they showed urea synthesis activity 7 days after treatment of FGF-4, HGF, oncostatin M, and dexamethasone. These results provided that HLSCs would be a useful cell source in the field of regenerative medicine as well as liver cell biology.
Product Description
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Urea, suitable for electrophoresis