Merck
  • Fusion of a xylan-binding module to gluco-oligosaccharide oxidase increases activity and promotes stable immobilization.
Fusion of a xylan-binding module to gluco-oligosaccharide oxidase increases activity and promotes stable immobilization.
PloS one (2014-04-17)
Thu V Vuong, Emma R Master
ABSTRACT
The xylan-binding module Clostridium thermocellum CBM22A was successfully fused to a gluco-oligosaccharide oxidase, GOOX-VN, from Sarocladium strictum via a short TP linker, allowing the fused protein to effectively bind different xylans. The presence of the CtCBM22A at the N-terminal of GOOX-VN increased catalytic activity on mono- and oligo-saccharides by 2-3 fold while not affecting binding affinity to these substrates. Notably, both GOOX-VN and its CBM fusion also showed oxidation of xylo-oligosaccharides with degrees of polymerization greater than six. Whereas fusion to CtCBM22A did not alter the thermostability of GOOX-VN or reduce substrate inhibition, CtCBM22A_GOOX-VN could be immobilized to insoluble oat spelt xylan while retaining wild-type activity. QCM-D analysis showed that the fused enzyme remained bound during oxidation. These features could be harnessed to generate hemicellulose-based biosensors that detect and quantify the presence of different oligosaccharides.
Product Description
Xylose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Xylose, ≥99%