Merck
  • Comparative evaluation of a new commercial colorimetric microdilution assay (SensiQuattro Candida EU) with MIC test strip and EUCAST broth microdilution methods for susceptibility testing of invasive Candida isolates.
Comparative evaluation of a new commercial colorimetric microdilution assay (SensiQuattro Candida EU) with MIC test strip and EUCAST broth microdilution methods for susceptibility testing of invasive Candida isolates.
Journal of clinical microbiology (2014-11-14)
Hedda Luise Koehling, Birgit Willinger, Jan Buer, Peter-Michael Rath, Joerg Steinmann
ABSTRACT
Candidemia is an important cause of morbidity and mortality in immunosuppressed patients. Candida isolates must be cultivated, identified, and tested for susceptibility. We compared the performance of a new colorimetric broth microdilution panel (SensiQuattro Candida EU) for antifungal susceptibility testing to that of Liofilchem's MIC test strip and the EUCAST reference broth microdilution protocol. We tested 187 blood culture isolates of 5 Candida spp. (120 C. albicans, 38 C. glabrata, 10 C. parapsilosis, 12 C. tropicalis, and 7 C. krusei) against seven antifungal agents (amphotericin B, fluconazole, voriconazole, posaconazole, caspofungin, anidulafungin, and micafungin) and interpreted the MICs according to the EUCAST recommendations. If applicable, the overall essential agreement (EA) of the SensiQuattro panel with the reference broth microdilution was slightly higher for C. albicans (87%) than for other species (85.8%). We found that SensiQuattro performed best in testing amphotericin B (EA, 100%), voriconazole (EA, 93.7%), and posaconazole (EA, 94.8%) against C. albicans, but its error rate for this species was high (29.6%) because of mainly major errors (26.7%) in testing anidulafungin and micafungin. Compared to the SensiQuattro panel, the MIC test strip exhibited a higher level of agreement for most isolates. SensiQuattro assays are easy to perform, but they are currently not suitable for testing echinocandins against Candida spp.
Product Description
Supelco
D-(+)-Glucose, analytical standard
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Sigma-Aldrich
Voriconazole, ≥98% (HPLC)
Supelco
Voriconazole, VETRANAL®, analytical standard
USP
Fluconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Voriconazole, European Pharmacopoeia (EP) Reference Standard
SAFC
MOPS
Fluconazole, European Pharmacopoeia (EP) Reference Standard
Flucytosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
5-Fluorocytosine, nucleoside analog
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Supelco
Posaconazole, VETRANAL®, analytical standard
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Flucytosine for system suitability, European Pharmacopoeia (EP) Reference Standard