Merck
  • Inhalation administration of valerena-4,7(11)-diene from Nardostachys chinensis roots ameliorates restraint stress-induced changes in murine behavior and stress-related factors.
Inhalation administration of valerena-4,7(11)-diene from Nardostachys chinensis roots ameliorates restraint stress-induced changes in murine behavior and stress-related factors.
Biological & pharmaceutical bulletin (2014-06-03)
Hiroaki Takemoto, Yuka Omameuda, Michiho Ito, Tatsuo Fukuda, Shuji Kaneko, Akinori Akaike, Yoshinori Kobayashi
ABSTRACT
Dried Nardostachys chinensis roots contain sesquiterpenoids that are widely used as herbal tranquilizers. We previously identified the highly sedative sesquiterpenoid valerena-4,7(11)-diene (VLD) from this plant. In the present study, we investigated stress reducing effects of VLD and the associated mechanisms of action. Application of 15-min restraint stresses induced excitatory behaviors in mice. Immobility times in the forced swim test and sleeping times in the pentobarbital sleep test were shortened in the stressed group by 47% and 43%, respectively, compared with the control group. Furthermore, restraint stress increased serum corticosterone levels by 75%, and cerebral serotonin (5-HT) and dopamine (DA) levels. Inhaled VLD (300 µg/cage) suppressed stress-induced excitatory behaviors and significantly reduced stress-induced blood corticosterone, cerebral 5-HT, and DA levels. These results suggest that VLD interacts with the hypothalamic-pituitary-adrenal axis and the sympathetic-adrenomedullary system. These interactions appear to involve GABAergic and D2 antagonist activities. Moreover, tests in anosmic and intravenously treated mice showed that the sedative effect of inhaled VLD was expressed via olfactory stimulation and pulmonary absorption. Although more studies are required to further elucidate the properties of this compound, our studies suggest that VLD may be an effective anti-stress aromatherapy for humans.
Product Description
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Corticosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diazepam
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Supelco
Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Supelco
Chlorpromazine hydrochloride, VETRANAL®, analytical standard
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
USP
Chlorpromazine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, BioXtra
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Caffeine, European Pharmacopoeia (EP) Reference Standard