Merck
Hydrogen sulfide inhibits Na+ uptake in larval zebrafish, Danio rerio.
Pflugers Archiv : European journal of physiology (2014-06-19)
Yusuke Kumai, Cosima S Porteus, Raymond W M Kwong, Steve F Perry
ABSTRACT
The present study investigated the role of hydrogen sulfide (H2S) in regulating Na(+) uptake in larval zebrafish, Danio rerio. Waterborne treatment of larvae at 4 days post-fertilization (dpf) with Na2S or GYY-4137 (chemicals known to generate H2S) significantly reduced Na(+) uptake. Exposure of larvae to water enriched with NaCl (1 mM NaCl) caused a pronounced reduction in Na(+) uptake which was prevented by pharmacological inhibition of cystathionine β-synthase (CBS) or cystathionine γ-lyase (CSE), two key enzymes involved in the endogenous synthesis of H2S. Furthermore, translational gene knockdown of CSE and CBSb significantly increased the basal rate of Na(+) uptake. Waterborne treatment with Na2S significantly decreased whole-body acid excretion and reduced Na(+) uptake in larval zebrafish preexposed to acidic (pH 4.0) water (a condition shown to promote Na(+) uptake via Na(+)-H(+)-exchanger 3b, NHE3b). However, Na2S did not affect Na(+) uptake in larvae depleted of NHE3b-containing ionocytes (HR cells) after knockdown of transcription factor glial cell missing 2 (gcm2) in which Na(+) uptake occurs predominantly via Na(+)-Cl(-) co-transporter (NCC)-containing cells. These observations suggest that Na(+) uptake via NHE3b, but not NCC, is regulated by H2S. Whole-mount immunohistochemistry demonstrated that ionocytes expressing NHE3b also express CSE. These data suggests a physiologically relevant role of H2S as a mechanism to lower Na(+) uptake in zebrafish larvae, probably through its inhibitory action on NHE3b.
Sigma-Aldrich
Hydrogen sulfide, ≥99.5%
Sigma-Aldrich
Sulfur-32S, 99.9 atom % 32S
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sigma-Aldrich
Hydrogen sulfide solution, 0.8 M in THF