Merck
  • Effect of adjuvant capecitabine or fluorouracil, with or without oxaliplatin, on survival outcomes in stage III colon cancer and the effect of oxaliplatin on post-relapse survival: a pooled analysis of individual patient data from four randomised controlled trials.
Effect of adjuvant capecitabine or fluorouracil, with or without oxaliplatin, on survival outcomes in stage III colon cancer and the effect of oxaliplatin on post-relapse survival: a pooled analysis of individual patient data from four randomised controlled trials.
The Lancet. Oncology (2014-12-03)
Hans-Joachim Schmoll, Chris Twelves, Weijing Sun, Michael J O'Connell, Thomas Cartwright, Edward McKenna, Muhammad Saif, Steve Lee, Greg Yothers, Daniel Haller
ABSTRACT
Oxaliplatin-based adjuvant therapy is the standard of care for stage III colon cancer. Adjuvant capecitabine with or without oxaliplatin versus leucovorin and fluorouracil with or without oxaliplatin has not been directly compared; therefore, we aimed to analyse the efficacy and safety of these treatments using individual patient data pooled from four randomised controlled trials. We also assessed post-relapse survival, which has been postulated to be worse in patients receiving adjuvant oxaliplatin. Patients with resected stage III colon cancer who were 18 years of age or older, with an Eastern Cooperative Oncology Group performance status of 0 or 1, from four randomised controlled trials (NSABP C-08, XELOXA, X-ACT, and AVANT; 8734 patients in total) were pooled and analysed. The treatment regimens included in our analyses were: XELOX (oxaliplatin and capecitabine); leucovorin and fluorouracil; capecitabine; FOLFOX-4 (leucovorin, fluorouracil, and oxaliplatin); and modified FOLFOX-6 (mFOLFOX-6). Disease-free survival was the primary endpoint for all trials that supplied patients for this analysis. Here, we compared disease-free, relapse-free, and overall survival between the patient groups who received capecitabine with or without oxaliplatin and those who received leucovorin and fluorouracil with or without oxaliplatin. Post-relapse survival was compared between the combined XELOX and FOLFOX groups, and the leucovorin and fluorouracil groups. Post-relapse survival was also compared between the capecitabine with or without oxaliplatin and leucovorin and fluorouracil with or without oxaliplatin groups. Disease-free survival did not differ significantly between patients who received leucovorin and fluorouracil versus those who received capecitabine in adjusted analyses (hazard ratio [HR] 1·02 [0·93-1·11; p=0·72]) or in unadjusted analyses (HR 1·01 [95% CI 0·92-1·10; p=0·86]). Relapse-free survival was similar (adjusted HR 1·02 [0·93-1·12; p=0·72] and unadjusted HR 1·01 [95% CI 0·92-1·11; p=0·86]), as was overall survival (adjusted HR 1·04 [95% CI 0·93-1·15; p=0·50] and unadjusted HR 1·02 [0·92-1·14]; p=0·65). For overall survival, a significant interaction between oxaliplatin and fluoropyrimidine was recorded in the multiple Cox regression analysis (p=0·014). Post-relapse survival was similar in adjusted (p=0·23) and unadjusted analyses (p=0·33) for the comparison of XELOX or FOLFOX versus leucovorin and fluorouracil, and was also similar for capecitabine-based regimens versus leucovorin and fluorouracil-based regimens (unadjusted p=0·26). Combination therapy with oxaliplatin provided consistently improved outcomes without adversely affecting post-relapse survival in the adjuvant treatment of stage III colon cancer, irrespective of whether the fluoropyrimidine backbone was capecitabine or leucovorin and fluorouracil. These data add to the existing evidence that oxaliplatin plus capecitabine or leucovorin and fluorouracil is the standard of care for the adjuvant treatment of stage III colon cancer, and offers physicians flexibility to treat patients according to the patients' overall physical performance and preference. Genentech Inc.
Product Description
Supelco
5-Fluorouracil, analytical standard
Capecitabine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Capecitabine, ≥98% (HPLC)
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oxaliplatin, powder
Sigma-Aldrich
2′-Deoxycytidine, ≥99% (HPLC)
USP
Oxaliplatin, United States Pharmacopeia (USP) Reference Standard
Supelco
Oxaliplatin, Pharmaceutical Secondary Standard; Certified Reference Material