Merck
Ultrasonic monitoring of malolactic fermentation in red wines.
Ultrasonics (2014-05-06)
D Novoa-Díaz, J M Rodríguez-Nogales, E Fernández-Fernández, J Vila-Crespo, J García-Álvarez, M A Amer, J A Chávez, A Turó, M J García-Hernández, J Salazar
ABSTRACT
The progress of malolactic fermentation in red wines has been monitored by using ultrasonic techniques. The evolution of ultrasonic velocity of a tone burst 1MHz longitudinal wave was measured, analyzed and compared to those parameters of oenological interest obtained simultaneously by analytical methods. Semi-industrial tanks were used during measurements pretending to be in real industrial conditions. Results showed that the ultrasonic velocity mainly changes as a result of the conversion by lactic acid bacteria of malic acid into lactic acid and CO2. Overall, the present study has demonstrated the potential of the ultrasonic technique in monitoring the malolactic fermentation process.
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Sigma-Aldrich
DL-Malic acid, ≥98% (capillary GC)
Sigma-Aldrich
D-(+)-Malic acid, unnatural form, ≥97.0% (T)
Sigma-Aldrich
Malic acid, meets USP/NF testing specifications
Supelco
D-Malic acid, analytical standard
Sigma-Aldrich
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
USP
Lactic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
DL-Malic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
DL-Malic acid, 99%
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Supelco
Malic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
DL-Malic acid, analytical standard
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
L-(−)-Malic acid disodium salt, ≥95% (titration)
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material