Merck
  • Similar effects of diets rich in stearic acid or trans-fatty acids on platelet function and endothelial prostacyclin production in humans.
Similar effects of diets rich in stearic acid or trans-fatty acids on platelet function and endothelial prostacyclin production in humans.
Arteriosclerosis, thrombosis, and vascular biology (1998-03-04)
A M Turpeinen, J Wübert, A Aro, R Lorenz, M Mutanen
ABSTRACT
The effects of stearic acid (C18:0) and trans-fatty acids (trans-FAs) on measures of platelet function and prostacyclin (PGI2) production are poorly understood in humans. In this controlled dietary study, platelet function and endothelial PGI2 production were studied in healthy humans after they consumed diets rich in C18:0 or trans-FAs. For 5 weeks, 80 subjects consumed a baseline diet high in saturated FAs and were then switched to a diet containing 9.3% of energy as stearic acid or a diet containing 8.7 energy% as trans-FAs from hydrogenated vegetable oils for another 5 weeks. All diets contained 32.2 to 33.9 energy% fat, 14.6 to 15.8 energy% saturated plus trans-FAs, 12.2 to 12.5 energy% cis-monounsaturated, and 2.9 to 3.5 energy% polyunsaturated FAs. No significant differences between the C18:0 and trans-FA diets were found in the urinary excretion of 2,3-dinor-thromboxane B2 or 2,3-dinor-6-keto-prostaglandin F1alpha. In vitro production of thromboxane B2 by platelets as well as urinary excretion of beta-thromboglobulin were also similar after both diets. Collagen-induced in vitro aggregation was significantly enhanced after the C18:0 diet compared with the trans-FA diet (P=.02), whereas no differences between the diets were found with ADP. The results indicate similar effects of C18:0 and trans-FA diets on platelet activation and endothelial PGI2 production.
Product Description
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard