Merck
  • Telmisartan/Hydrochlorothiazide combination therapy for the treatment of hypertension: a pooled analysis in older and younger patients.
Telmisartan/Hydrochlorothiazide combination therapy for the treatment of hypertension: a pooled analysis in older and younger patients.
Journal of clinical hypertension (Greenwich, Conn.) (2013-06-05)
Sverre E Kjeldsen, Helmut Schumacher, Steen Neldam, Robert M Guthrie
ABSTRACT
Older patients frequently receive angiotensin II receptor blocker/diuretic combinations to control blood pressure (BP), although there have been relatively few trials specifically examining this patient population. A pooled analysis was performed of data from 7 randomized trials of telmisartan/hydrochlorothiazide combinations or telmisartan monotherapy in older (65 years and older) and younger (younger than 65 years) patients to better understand the response of older patients to a telmisartan/hydrochlorothiazide combination. Telmisartan doses were 40 mg and 80 mg (T40 and T80). Hydrochlorothiazide doses were 12.5 mg and 25 mg (H12.5 and H25). A total of 3654 patients were included and the mean treatment duration was approximately 8 weeks. BP reductions with telmisartan/hydrochlorothiazide combinations were broadly similar in older and younger patients. In older patients, mean BP reductions from baseline were -30.1/-19.0 mm Hg with the T80/H25 combination and -21.7/-13.0 mm Hg with T80 monotherapy. Tolerability was similar regardless of age, and the incidence of adverse events in both older and younger patients was similar to placebo. The telmisartan/hydrochlorothiazide combination, particularly high-dose T80/H25, is effective and well tolerated in patients 65 years and older as well as in younger patients.
Product Description
Supelco
Hydrochlorothiazide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Hydrochlorothiazide, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydrochlorothiazide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrochlorothiazide, meets USP testing specifications
Sigma-Aldrich
Hydrochlorothiazide, crystalline
Hydrochlorothiazide, European Pharmacopoeia (EP) Reference Standard
Hydrochlorothiazide for peak identification, European Pharmacopoeia (EP) Reference Standard