Merck
  • Coenzyme Q10 depletion in medical and neuropsychiatric disorders: potential repercussions and therapeutic implications.
Coenzyme Q10 depletion in medical and neuropsychiatric disorders: potential repercussions and therapeutic implications.
Molecular neurobiology (2013-06-14)
Gerwyn Morris, George Anderson, Michael Berk, Michael Maes
ABSTRACT
Coenzyme Q10 (CoQ10) is an antioxidant, a membrane stabilizer, and a vital cofactor in the mitochondrial electron transport chain, enabling the generation of adenosine triphosphate. It additionally regulates gene expression and apoptosis; is an essential cofactor of uncoupling proteins; and has anti-inflammatory, redox modulatory, and neuroprotective effects. This paper reviews the known physiological role of CoQ10 in cellular metabolism, cell death, differentiation and gene regulation, and examines the potential repercussions of CoQ10 depletion including its role in illnesses such as Parkinson's disease, depression, myalgic encephalomyelitis/chronic fatigue syndrome, and fibromyalgia. CoQ10 depletion may play a role in the pathophysiology of these disorders by modulating cellular processes including hydrogen peroxide formation, gene regulation, cytoprotection, bioenegetic performance, and regulation of cellular metabolism. CoQ10 treatment improves quality of life in patients with Parkinson's disease and may play a role in delaying the progression of that disorder. Administration of CoQ10 has antidepressive effects. CoQ10 treatment significantly reduces fatigue and improves ergonomic performance during exercise and thus may have potential in alleviating the exercise intolerance and exhaustion displayed by people with myalgic encepholamyletis/chronic fatigue syndrome. Administration of CoQ10 improves hyperalgesia and quality of life in patients with fibromyalgia. The evidence base for the effectiveness of treatment with CoQ10 may be explained via its ability to ameliorate oxidative stress and protect mitochondria.
Product Description
Ubidecarenone for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Ubidecarenone, United States Pharmacopeia (USP) Reference Standard
USP
Ubidecarenone for System Suitability, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Coenzyme Q10, ≥98% (HPLC)
Supelco
Coenzyme Q10, analytical standard
Ubidecarenone, European Pharmacopoeia (EP) Reference Standard