Merck
  • Topical capsaicin (high concentration) for chronic neuropathic pain in adults.
Topical capsaicin (high concentration) for chronic neuropathic pain in adults.
The Cochrane database of systematic reviews (2013-03-02)
Sheena Derry, Andrew Sven-Rice, Peter Cole, Toni Tan, R Andrew Moore
ABSTRACT
Topical creams with capsaicin are used to treat peripheral neuropathic pain. Following application to the skin capsaicin causes enhanced sensitivity, followed by a period with reduced sensitivity and, after repeated applications, persistent desensitisation. High-concentration (8%) capsaicin patches were developed to increase the amount of capsaicin delivered; rapid delivery was thought to improve tolerability because cutaneous nociceptors are 'defunctionalised' quickly. The single application avoids noncompliance. Only the 8% patch formulation of capsaicin is available, with a capsaicin concentration about 100 times greater than conventional creams.High-concentration topical capsaicin is given as a single patch application to the affected part. It must be applied under highly controlled conditions, normally under local anaesthetic, due to the initial intense burning sensation it causes. The benefits are expected to last for about 12 weeks, when another application might be made. To review the evidence from controlled trials on the efficacy and tolerability of topically applied, high-concentration (8%) capsaicin in chronic neuropathic pain in adults. We searched CENTRAL, MEDLINE, EMBASE and clinicaltrials.gov to December 2012. Randomised, double-blind, placebo-controlled studies of at least six weeks' duration, using topical capsaicin to treat neuropathic pain. Two review authors independently assessed trial quality and validity, and extracted data on numbers of participants with pain relief (clinical improvement) after at least six weeks, and with local skin reactions. We calculated risk ratio and numbers needed to treat to benefit (NNT) and harm (NNH). We sought details of definition of pain relief and specific adverse events.Efficacy outcomes reflecting long-duration pain relief after a single drug application were from the patient global impression of change (PGIC) at specific points, usually eight and 12 weeks. We regarded these outcomes as first-tier evidence. We regarded average pain scores over weeks 2 to 8 and 2 to 12 and the number and/or percentage of participants with pain intensity reduction of at least 30% or at least 50% over baseline as second-tier evidence. We included six studies, involving 2073 participants; they were of generally good reporting quality; the control was 0.04% topical capsaicin to help maintain blinding. Efficacy outcomes were inconsistently reported between studies, however, resulting in analyses for most outcomes being based on less than complete data.Four studies involved 1272 participants with postherpetic neuralgia. All efficacy outcomes were significantly better than control. At both eight and 12 weeks there was a significant benefit for high-concentration over low-concentration topical capsaicin for participants reporting themselves to be much or very much better, with point estimates of the NNTs of 8.8 (95% confidence interval (CI) 5.3 to 26) and 7.0 (95% CI 4.6 to 15) respectively. More participants had average 2 to 8-week and 2 to 12-week pain intensity reductions over baseline of at least 30% and at least 50% with active treatment than control, with NNT values between 10 and 12.Two studies involved 801 participants with painful HIV-neuropathy. In a single study the NNT at 12 weeks for participants to be much or very much better was 5.8 (95% CI 3.8 to 12). Over both studies more participants had average 2 to 12-week pain intensity reductions over baseline of at least 30% with active treatment than control, with an NNT of 11.Local adverse events were common, but not consistently reported. Serious adverse events were no more common with active treatment (4.1%) than control (3.2%). Adverse event withdrawals did not differ between groups, but lack of efficacy withdrawals were somewhat more common with control than active treatment, based on small numbers of events. No deaths were judged to be related to study medication. High-concentration topical capsaicin used to treat postherpetic neuralgia and HIV-neuropathy generates more participants with high levels of pain relief than does control treatment using a much lower concentration of capsaicin. The additional proportion who benefit over control is not large, but for those who do obtain high levels of pain relief there are additional improvements in sleep, fatigue, depression and an improved quality of life. High-concentration topical capsaicin is therefore similar to other therapies for chronic pain. In this case, the high cost of single and repeated applications suggest that high-concentration topical capsaicin is likely to be used when other available therapies have failed, and that it should probably not be used repeatedly without substantial documented pain relief. Even when efficacy is established, there are unknown risks, especially on epidermal innervation, of repeated application of long periods.
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Supelco
Capsaicin, analytical standard
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
Capsaicin, natural
USP
Capsaicin, United States Pharmacopeia (USP) Reference Standard
Capsaicin, European Pharmacopoeia (EP) Reference Standard