Merck
  • Farnesol-induced cell death in the filamentous fungus Aspergillus nidulans.
Farnesol-induced cell death in the filamentous fungus Aspergillus nidulans.
Biochemical Society transactions (2011-09-23)
Taísa Magnani Dinamarco, Maria Helena S Goldman, Gustavo Henrique Goldman
ABSTRACT
FOH (farnesol), a non-sterol isoprenoid produced by dephosphorylation of farnesyl pyrophosphate, has been shown to inhibit proliferation and induce apoptosis. We have been using Aspergillus nidulans and FOH as a model system and cell death stimulus, respectively, aiming to understand by which means filamentous fungi are driven towards cell death. Here, we review some of our findings about FOH-induced cell death in A. nidulans.
Sigma-Aldrich
Farnesol, mixture of isomers, ≥95%, stabilized, FG
Sigma-Aldrich
Farnesol, 95%
Supelco
Farnesol, analytical standard, stabilized, mixture of isomers