Merck
  • 1-Methylimidazolium hydrogen sulfate catalyzed convenient synthesis of 2,5-dimethyl-N-substituted pyrroles under ultrasonic irradiation.
1-Methylimidazolium hydrogen sulfate catalyzed convenient synthesis of 2,5-dimethyl-N-substituted pyrroles under ultrasonic irradiation.
Ultrasonics sonochemistry (2013-03-15)
Daqing Li, Hongjun Zang, Changchun Wu, NianQin Yu
ABSTRACT
Ionic liquid [HMIM]HSO4 was found to be an efficient catalyst for the synthesis of N-substituted pyrroles through the reaction of 2,5-hexanedione with amines under ultrasonic irradiation at room temperature. These reactions proceed with good yields under short reaction time. Furthermore, the green catalytic system can be recycled specific times with no decreases in yields and reaction rates.
Sigma-Aldrich
1-Methylimidazolium hydrogen sulfate, 95%
Sigma-Aldrich
1-Methylimidazole, ReagentPlus®, 99%
Sigma-Aldrich
1-Methylimidazole, ≥99%, purified by redistillation