Merck
  • Pyelonephritis in slaughter pigs and sows: morphological characterization and aspects of pathogenesis and aetiology.
Pyelonephritis in slaughter pigs and sows: morphological characterization and aspects of pathogenesis and aetiology.
Acta veterinaria Scandinavica (2010-08-14)
Louise K Isling, Bent Aalbaek, Malene Schrøder, Páll S Leifsson
ABSTRACT
Pyelonephritis is a serious disease in pig production that needs to be further studied. The purpose of this study was to describe the morphology, investigate the pathogenesis, and evaluate the aetiological role of Escherichia coli in pyelonephritis in slaughtered pigs by concurrent bacteriological, gross and histopathological examinations. From Danish abattoirs, kidneys and corresponding lymph nodes from 22 slaughtered finishing pigs and 26 slaughtered sows with pyelonephritis were collected and evaluated by bacteriology and pathology. Based on gross lesions, each kidney (lesion) was grouped as acute, chronic, chronic active, or normal and their histological inflammatory stage was determined as normal (0), acute (1), sub-acute (2), chronic active (3), or chronic (4). Immunohistochemical identification of neutrophils, macrophages, T-lymphocytes, B-lymphocytes, plasma cells, E. coli and Tamm-Horsfall protein (THP) in renal sections was performed. The number of E. coli and the proportion of immunohistochemically visualized leukocytes out of the total number of infiltrating leukocytes were scored semi-quantitatively. Lesions in finishing pigs and sows were similar. Macroscopically, multiple unevenly distributed foci of inflammation mostly affecting the renal poles were observed. Histologically, tubulointerstitial infiltration with neutrophils and mononuclear cells and tubular destruction was the main findings. The significant highest scores of L1 antigen+ neutrophils were in inflammatory stage 1 while the significant highest scores of CD79alphacy+ B-lymphocytes, IgG+ and IgA+ plasma cells were in stage 3 or 4. Neutrophils were the dominant leukocytes in stage 1 while CD3epsilon+ T-lymphocytes dominated in stage 2, 3 and 4. Interstitially THP was seen in 82% and 98% of kidneys with pyelonephritis from finishing pigs and sows, respectively. E. coli was demonstrated in monoculture and/or identified by immunohistochemistry in relation to inflammation in four kidneys from finishing pigs and in 34 kidneys from sows. E. coli played a significant role in the aetiology of pyelonephritis. Neutrophils were involved in the first line of defence. CD3epsilon+ T-lymphocytes were involved in both the acute and chronic inflammatory response while a humoral immune response was most pronounced in later inflammatory stages. The observed renal lesions correspond with an ascending bacterial infection with presence of intra-renal reflux.
Sigma-Aldrich
IgG from goat serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution