Merck
Cyclooxygenase-1 haplotype modulates platelet response to aspirin.
Journal of thrombosis and haemostasis : JTH (2005-09-10)
A O Maree, R J Curtin, A Chubb, C Dolan, D Cox, J O'Brien, P Crean, D C Shields, D J Fitzgerald
ABSTRACT
Aspirin (acetylsalicylic acid) irreversibly inhibits platelet cyclooxygenase (COX)-1, the enzyme that converts arachidonic acid (AA) to the potent platelet agonist thromboxane (TX) A2. Despite clear benefit from aspirin in patients with cardiovascular disease (CAD), evidence of heterogeneity in the way individuals respond has given rise to the concept of 'aspirin resistance.' To evaluate the hypothesis that incomplete suppression of platelet COX as a consequence of variation in the COX-1 gene may affect aspirin response and thus contribute to aspirin resistance. Aspirin response, determined by serum TXB2 levels and AA-induced platelet aggregation, was prospectively studied in patients (n = 144) with stable CAD taking aspirin (75-300 mg). Patients were genotyped for five single nucleotide polymorphisms in COX-1 [A-842G, C22T (R8W), G128A (Q41Q), C644A (G213G) and C714A (L237M)]. Haplotype frequencies and effect of haplotype on two platelet phenotypes were estimated by maximum likelihood. The four most common haplotypes were considered separately and less common haplotypes pooled. COX-1 haplotype was significantly associated with aspirin response determined by AA-induced platelet aggregation (P = 0.004; 4 d.f.). Serum TXB2 generation was also related to genotype (P = 0.02; 4 d.f.). Genetic variability in COX-1 appears to modulate both AA-induced platelet aggregation and thromboxane generation. Heterogeneity in the way patients respond to aspirin may in part reflect variation in COX-1 genotype.
Product Description
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard