Merck
  • Model sclerotization studies. 4. Generation of N-acetylmethionyl catechol adducts during tyrosinase-catalyzed oxidation of catechols in the presence of N-acetylmethionine.
Model sclerotization studies. 4. Generation of N-acetylmethionyl catechol adducts during tyrosinase-catalyzed oxidation of catechols in the presence of N-acetylmethionine.
Archives of insect biochemistry and physiology (1998-05-20)
M Sugumaran, E Nelson
ABSTRACT
Incubation of catechol with mushroom tyrosinase in the presence of N-acetylmethionine resulted in the generation of an adduct. This product was identified to be N-acetylmethionyl catechol, on the basis of spectral characteristics and well-characterized chemical reaction of o-benzoquinone with N-acetylmethionine. Enzyme-catalyzed oxidation of catechol and the subsequent nonenzymatic addition of the resultant quinone to N-acetylmethionine accounted for the observed reaction. That the reaction is not confined to catechol alone, but is of general occurrence, can be demonstrated by the facile generation of similar adducts in incubation mixtures containing N-acetylmethionine, tyrosinase, and different N-acetylmethionines, such as 4-methylcatechol and N-acetyldopamine. Attempts to duplicate the reaction with insect cuticular phenoloxidases were not successful, as the excess N-acetylmethionine used in the reaction inhibited their activity. Nevertheless, occurrence of this nonenzymatic reactivity. Nevertheless, occurrence of this nonenzymatic reaction between N-acetylmethionine and mushroom tyrosinase-generated quinones indicates that a similar reaction between enzymatically generated quinones in the cuticle with protein-bound methionine moiety is likely to occur during in vivo quinone tanning as well.
Sigma-Aldrich
N-Acetyl-L-methionine, ≥98.5% (T)
Sigma-Aldrich
N-Acetyl-D-methionine, ~99%