Merck
  • Fit of fluxes of sunscreens and other compounds from propylene glycol:water (30:70) through human skin and silicone membrane to the Roberts-Sloan equation: the effect of polar vehicle (or water) solubility.
Fit of fluxes of sunscreens and other compounds from propylene glycol:water (30:70) through human skin and silicone membrane to the Roberts-Sloan equation: the effect of polar vehicle (or water) solubility.
Journal of cosmetic science (2013-06-12)
Kenneth B Sloan, Hemamalini Devarajan-Ketha, Jennifer Synovec, Susruta Majumdar
ABSTRACT
It would be useful to develop a surrogate for animal skin, which could be use to predict flux through human skin. The fluxes (and physicochemical properties) of sunscreens and other compounds from propylene glycol (PG):water (AQ), 30:70, through human skin have previously been reported. We measured the fluxes of several of those sunscreens and other compounds from PG:AQ, 30:70, through silicone membrane and fit both sets of data to the Roberts-Sloan (RS) equation to determine any similarities. For both sets of data, the fluxes were directly dependent on their solubilities in a lipid solvent [octanol (OCT), in this case] and in a polar solvent (PG:AQ, 30:70, or AQ in this case) and inversely on their molecular weights. The fit of the experimental (EXP) fluxes through human skin in vivo to RS was excellent: r² = 0.92 if the vehicle (VEH) PG:AQ, 30:70 was the polar solvent (RS¹) or r² = 0.97 if water was the polar solvent (RS²). The fit of the EXP fluxes through silicone membrane to RS was good: r² = 0.80 if the VEH PG:AQ, 30:70, was the polar solvent (RS¹) or r² = 0.81 if water was the polar solvent (RS²). The correlations between their EXP fluxes through human skin in vivo and their EXP fluxes through silicone membrane were good (r² = 0.85). In addition, the correlation between EXP fluxes from PG:AQ, 30:70, through human skin in vivo and their fluxes calculated from the coefficients of the fit of solubilities, molecular weights and fluxes from water through silicone membranes from a previous n = 22 database to RS was even better (r² = 0.94). These results suggest that flux through human skin can be calculated from flux through a silicone membrane.
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
(S)-(+)-1,2-Propanediol, 96%
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
(R)-(−)-1,2-Propanediol, 96%
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material