Merck
  • Inotropic and lusitropic effects of calcitonin gene-related peptide in the heart.
Inotropic and lusitropic effects of calcitonin gene-related peptide in the heart.
American journal of physiology. Heart and circulatory physiology (2013-04-16)
Mustafa Al-Rubaiee, Pandu R Gangula, Richard M Millis, Robin K Walker, Nsini A Umoh, Valerie M Cousins, Miara A Jeffress, Georges E Haddad
ABSTRACT
Previous studies have demonstrated positive-inotropic effects of calcitonin gene-related peptide (CGRP), but the mechanisms remain unclear. Therefore, two experiments were performed to determine the physiological correlates of the positive-inotropic effects of CGRP. Treatments designed to antagonize the effects of physiologically active CGRP₁₋₃₇ included posttreatment with CGRP₈₋₃₇ and pretreatment with LY-294002 (LY, an inhibitor of phosphatidylinositol 3-kinase), 17β-estradiol (E), and progesterone (P) were also used to modulate the effects of CGRP₁₋₃₇. Experiment 1 was in vitro studies on sarcomeres and cells of isolated adult rat cardiac myocytes. CGRP₁₋₃₇, alone and in combination with E and P, decreased sarcomere shortening velocities and increased shortening percentages, effects that were antagonized by CGRP₈₋₃₇, but not by LY. CGRP₁₋₃₇ increased resting intracellular calcium ion concentrations and Ca(2+) influxes, effects that were also antagonized by both CGRP₈₋₃₇ and LY. Experiment 2 was in vivo studies on left ventricular pressure-volume (PV) loops. CGRP₁₋₃₇ increased end-systolic pressure, ejection fraction, and velocities of contraction and relaxation while decreasing stroke volume, cardiac output, stroke work, PV area, and compliance. After partial occlusion of the vena cava, CGRP₁₋₃₇ increased the slope of the end-systolic PV relationship. CGRP₈₋₃₇ and LY attenuated most of the CGRP-induced changes. These findings suggest that CGRP-induced positive-inotropic effects may be increased by treatments with estradiol and progesterone and inhibited by LY. The physiological correlates of CGRP-induced positive inotropy observed in rat sarcomeres, cells, and intact hearts are likely to reveal novel mechanisms of heart failure in humans.
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, ≥99%
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®