Merck
  • Gold nanorod-facilitated localized heating of droplets in microfluidic chips.
Gold nanorod-facilitated localized heating of droplets in microfluidic chips.
Optics express (2013-02-08)
Zhiyong Li, Pan Wang, Limin Tong, Lei Zhang
ABSTRACT
A gold nanorod-facilitated optical heating method for droplets in microfluidic chips is reported. Individual and stream nanoliter level droplets containing gold nanorods are heated by a low power 808-nm-wavelength laser. Owing to the high photothermal conversion efficiency of gold nanorods, a droplet temperature of 95 °C is achieved by employing a 13.6 mW laser with good reproducibility. The heating and cooling times are 200 and 800 ms, respectively, which are attributed to the fast thermal-transfer rates of the droplets. By controlling the irradiation laser power, the temperature cycles for polymerase chain reaction are also demonstrated.
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Silicone oil, for melting point and boiling point apparatuses
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, for oil baths (from −50°C to +200°C)
Sigma-Aldrich
Silicone oil, viscosity 60,000 cSt (25 °C)