Merck
  • Resonance Raman and electrocatalytic behavior of thiolate and imidazole bound iron porphyrin complexes on self assembled monolayers: functional modeling of cytochrome P450.
Resonance Raman and electrocatalytic behavior of thiolate and imidazole bound iron porphyrin complexes on self assembled monolayers: functional modeling of cytochrome P450.
Inorganic chemistry (2013-01-30)
Kushal Sengupta, Sudipta Chatterjee, Subhra Samanta, Sabyasachi Bandyopadhyay, Abhishek Dey
ABSTRACT
Electrodes bearing thiolate and imidazole coordinated iron porphyrin catalysts are constructed and characterized using resonance Raman spectroscopy, absorption spectroscopy, and electrochemistry. The cyclic voltammetry data and their pH dependences are used to establish the nature of the exchangeable trans ligands in both of these cases. In situ monitoring of partially reduced oxygen species (PROS) produced during O(2) reduction using rotating ring disc electrochemistry (RRDE) experiments provide direct insight into the "push-effect" of the thiolate ligand. The thiolate bound iron porphyrin electrode generates highly oxidizing species on the electrode during electrocatalytic O(2) reductions which are very reactive. These surfaces can utilize these oxidants to catalytically hydroxylate strong C-H bonds using molecular O(2) with turnover numbers as high as 200.
Sigma-Aldrich
Imidazole sodium derivative, technical grade
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole hydrochloride
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Imidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O