Merck
  • o-Benzenedisulfonimido-sulfonamides are potent inhibitors of the tumor-associated carbonic anhydrase isoforms CA IX and CA XII.
o-Benzenedisulfonimido-sulfonamides are potent inhibitors of the tumor-associated carbonic anhydrase isoforms CA IX and CA XII.
Bioorganic & medicinal chemistry (2013-01-29)
Özlen Güzel-Akdemir, Atilla Akdemir, Semra Isik, Daniela Vullo, Claudiu T Supuran
ABSTRACT
By using phthalimido-substituted aromatic sufonamides as lead molecules, a series of new sulfonamides incorporating ortho-benzenedisulfonimide moieties have been synthesized and tested against the human (h) cytosolic carbonic anhydrase (CA, EC 4.2.1.1) isozymes hCA I and hCA II and the transmembrane, tumor-associated isozymes hCA IX and hCA XII. All these compounds showed Ki values lower than 100nM and many of them showed better Kis than the reference compound acetazolamide, a clinically used sulfonamide. The tumor-associated isozymes were better inhibited than the cytosolic ones. A molecular docking within the active site of some CA isoforms, such as hCA I, explained these findings, as the benzenedisulfonimide moiety makes favorable interactions (hydrogen bonds) with amino acid residues involved in binding of inhibitors, such as Gln92, His67, and His64.
Product Description
Acetazolamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetazolamide, ≥99%, powder