Merck
  • Gold-catalyzed domino cycloisomerization/Pictet-Spengler reaction of 2-(4-aminobut-1-yn-1-yl)anilines with aldehydes: synthesis of tetrahydropyrido[4,3-b]indole scaffolds.
Gold-catalyzed domino cycloisomerization/Pictet-Spengler reaction of 2-(4-aminobut-1-yn-1-yl)anilines with aldehydes: synthesis of tetrahydropyrido[4,3-b]indole scaffolds.
The Journal of organic chemistry (2012-12-05)
B V Subba Reddy, Manisha Swain, S Madhusudana Reddy, J S Yadav, B Sridhar
ABSTRACT
A domino cycloisomerization/Pictet-Spengler reaction of 2-(4-aminobut-1-yn-1-yl)aniline with aldehydes has been achieved using a AuIPrCl (5 mol %)/AgSbF(6) (10 mol %) catalytic system to produce the corresponding 1-aryl-N-tosyl-2,3,4,5-tetrahydropyrido[4,3-b] indole derivatives in good yields. This is the first report on the synthesis of tetrahydro pyrido[4,3-b]indole scaffolds through as tandem 5-endo-dig cyclization and Pictect-Spengler reaction.
Product Description
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Sigma-Aldrich
Aniline hydrochloride, ≥99%