Merck
  • Facile synthesis of 4-substituted 3,4-dihydrocoumarins via an organocatalytic double decarboxylation process.
Facile synthesis of 4-substituted 3,4-dihydrocoumarins via an organocatalytic double decarboxylation process.
Organic & biomolecular chemistry (2012-03-01)
Shiyong Peng, Lei Wang, Haibing Guo, Shaofa Sun, Jian Wang
ABSTRACT
3,4-Dihydrocoumarins, considered to be valuable building blocks, have attracted considerable attention due to their various biological activities. Herein, we have documented an efficient and convenient double decarboxylation process for the synthesis of 4-substituted 3,4-dihydrocoumarin in moderate to excellent yields under mild reaction conditions (up to 98%).
Sigma-Aldrich
Dihydrocoumarin, 99%
Sigma-Aldrich
Dihydrocoumarin, ≥99%, FCC, FG