Merck
Immediate-type hypersensitivity to succinylated corticosteroids.
International archives of allergy and immunology (2010-11-27)
Annett Isabel Walker, Helen-Caroline Räwer, Wolfgang Sieber, Bernhard Przybilla
ABSTRACT
Despite their frequent use, systemic corticosteroids have rarely elicited immediate-type reactions. We report two male patients, aged 26 and 70 years, respectively, with severe immediate-type hypersensitivity secondary to the administration of corticosteroids esterified with succinate. Skin tests, basophil activation tests and challenge tests were performed for diagnostic evaluation. In both patients, immediate-type skin test reactions were found to methylprednisolone sodium hemisuccinate (MSH) and prednisolone sodium hemisuccinate (PSH). In contrast, nonsuccinylated corticosteroids (including methylprednisolone and prednisolone in one patient) yielded no test reactions. Basophils from one patient exhibited a stimulated expression of the activation marker CD63 upon in vitro incubation with PSH or hydrocortisone sodium succinate, but not with hydrocortisone. Skin tests and basophil activation tests were negative in controls. One patient was challenged with the incriminated drugs. He developed flush, conjunctivitis, tachycardia and dyspnea 2 min after injection of MSH, and dyspnea shortly after intravenous administration of PSH. Oral and intravenous challenge tests with nonsuccinylated corticosteroids were tolerated well by both patients. These case reports should alert clinicians to rare, but severe immediate-type reactions to corticosteroids, related to the succinate moiety in our patients. In case of allergic reactions to corticosteroids, it is mandatory to identify the causative agent and find safe alternatives.
Product Description
Supelco
Hydrocortisone 21-hemisuccinate, analytical standard
Sigma-Aldrich
Prednisolone 21-hemisuccinate sodium salt, ≥90%, powder
Sigma-Aldrich
Hydrocortisone 21-hemisuccinate sodium salt, ≥90% (HPLC)
Sigma-Aldrich
Hydrocortisone 21-hemisuccinate sodium salt, powder, BioReagent, suitable for cell culture