Merck
  • Isotopomer encapsulation in a cylindrical molecular capsule: a probe for understanding noncovalent isotope effects on a molecular level.
Isotopomer encapsulation in a cylindrical molecular capsule: a probe for understanding noncovalent isotope effects on a molecular level.
Journal of the American Chemical Society (2004-06-24)
Dalit Rechavi, Alessandro Scarso, Julius Rebek
ABSTRACT
Isotope effects for the encapsulation of deuterated versus nondeuterated guests are determined. This system involves the use of social isomers, and the origin of the isotope effect is localized to methyl groups interacting with aromatic rings.
Sigma-Aldrich
Mesitylene, reagent grade, 97%
Sigma-Aldrich
Mesitylene, 98%
Supelco
Mesitylene, analytical standard