Merck
  • Systemic delivery of AAV-GCDH ameliorates HLD-induced phenotype in a glutaric aciduria type I mouse model.
Systemic delivery of AAV-GCDH ameliorates HLD-induced phenotype in a glutaric aciduria type I mouse model.
Molecular therapy. Methods & clinical development (2024-07-10)
Anna Mateu-Bosch, Eulàlia Segur-Bailach, Emma Muñoz-Moreno, María José Barallobre, Maria Lourdes Arbonés, Sabrina Gea-Sorlí, Frederic Tort, Antonia Ribes, Judit García-Villoria, Cristina Fillat
ABSTRACT
Glutaric aciduria type 1 (GA1) is a rare inherited metabolic disorder caused by a deficiency of glutaryl-coenzyme A dehydrogenase (GCDH), with accumulation of neurotoxic metabolites, resulting in a complex movement disorder, irreversible brain damage, and premature death in untreated individuals. While early diagnosis and a lysine restricted diet can extend survival, they do not prevent neurological damage in approximately one-third of treated patients, and more effective therapies are required. Here we report the efficacy of adeno-associated virus 9 (AAV9)-mediated systemic delivery of human GCDH at preventing a high lysine diet (HLD)-induced phenotype in Gcdh -/- mice. Neonatal treatment with AAV-GCDH restores GCDH expression and enzyme activity in liver and striatum. This treatment protects the mice from HLD-aggressive phenotype with all mice surviving this exposure; in stark contrast, a lack of treatment on an HLD triggers very high accumulation of glutaric acid, 3-hydroxyglutaric acid, and glutarylcarnitine in tissues, with about 60% death due to brain accumulation of toxic lysine metabolites. AAV-GCDH significantly ameliorates the striatal neuropathology, minimizing neuronal dysfunction, gliosis, and alterations in myelination. Magnetic resonance imaging findings show protection against striatal injury. Altogether, these results provide preclinical evidence to support AAV-GCDH gene therapy for GA1.
Sigma-Aldrich
Anti-GCDH antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-GAPDH Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid