Merck
  • Mitochondrial metabolism sustains CD8+ T cell migration for an efficient infiltration into solid tumors.
Mitochondrial metabolism sustains CD8+ T cell migration for an efficient infiltration into solid tumors.
Nature communications (2024-03-12)
Luca Simula, Mattia Fumagalli, Lene Vimeux, Irena Rajnpreht, Philippe Icard, Gary Birsen, Dongjie An, Frédéric Pendino, Adrien Rouault, Nadège Bercovici, Diane Damotte, Audrey Lupo-Mansuet, Marco Alifano, Marie-Clotilde Alves-Guerra, Emmanuel Donnadieu
ABSTRACT
The ability of CD8+ T cells to infiltrate solid tumors and reach cancer cells is associated with improved patient survival and responses to immunotherapy. Thus, identifying the factors controlling T cell migration in tumors is critical, so that strategies to intervene on these targets can be developed. Although interstitial motility is a highly energy-demanding process, the metabolic requirements of CD8+ T cells migrating in a 3D environment remain unclear. Here, we demonstrate that the tricarboxylic acid (TCA) cycle is the main metabolic pathway sustaining human CD8+ T cell motility in 3D collagen gels and tumor slices while glycolysis plays a more minor role. Using pharmacological and genetic approaches, we report that CD8+ T cell migration depends on the mitochondrial oxidation of glucose and glutamine, but not fatty acids, and both ATP and ROS produced by mitochondria are required for T cells to migrate. Pharmacological interventions to increase mitochondrial activity improve CD8+ T cell intratumoral migration and CAR T cell recruitment into tumor islets leading to better control of tumor growth in human xenograft models. Our study highlights the rationale of targeting mitochondrial metabolism to enhance the migration and antitumor efficacy of CAR T cells in treating solid tumors.
Sigma-Aldrich
Oligomycin, A mixture of A, B, and C isomers.
Sigma-Aldrich
Etomoxir, A cell-permeable, irreversible, and stereospecific compound that is shown to inhibit carnitine palmitoyltransferase (CPT)-1 and DGAT activity in the mitochondria of rat heart H9c2 myoblastic cells.
Sigma-Aldrich
Glutaminase Inhibitor, Compound 968, Glutaminase Inhibitor, Compd 968, is a cell-permeable, reversible inhibitor of mitochondrial glutaminase. Represses growth & invasive activity in glutaminase upregulated fibroblasts and tumor cells.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
3-Nitropropionic acid, ≥97%
Sigma-Aldrich
Anti-OGDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)
Sigma-Aldrich
PS10, ≥98% (HPLC)