Merck
  • Breaking the Base Barrier: An Electron-Deficient Palladium Catalyst Enables the Use of a Common Soluble Base in C-N Coupling.
Breaking the Base Barrier: An Electron-Deficient Palladium Catalyst Enables the Use of a Common Soluble Base in C-N Coupling.
Journal of the American Chemical Society (2018-03-13)
Joseph M Dennis, Nicholas A White, Richard Y Liu, Stephen L Buchwald
ABSTRACT
Due to the low intrinsic acidity of amines, palladium-catalyzed C-N cross-coupling has been plagued continuously by the necessity to employ strong, inorganic, or insoluble bases. To surmount the many practical obstacles associated with these reagents, we utilized a commercially available dialkyl triarylmonophosphine-supported palladium catalyst that facilitates a broad range of C-N coupling reactions in the presence of weak, soluble bases. The mild and general reaction conditions show extraordinary tolerance for even highly base-sensitive functional groups. Additionally, insightful heteronuclear NMR studies using 15N-labeled amine complexes provide evidence for the key acidifying effect of the cationic palladium center.
Sigma-Aldrich
AdBrettPhos, 95%
Sigma-Aldrich
AdBrettPhos ChemBeads