Merck
Comparative retinal morphology of the platypus.
Journal of morphology (2011-05-14)
Caroline J Zeiss, Ivan R Schwab, Christopher J Murphy, Richard W Dubielzig
ABSTRACT
The purpose of this study is to identify evolutionary origin and fate of anatomic features of the duck-billed platypus eye. Eyes from the duck-billed platypus and four key evolutionary basal vertebrates (Pacific hagfish, north hemisphere sea lamprey, and Australian and South American lungfishes) were prepared for light microscopy. In addition to a standard panel of stains, tissues were immunostained against a variety of rod and cone opsins. Finally, published opsin sequences of platypus and several other vertebrate species were aligned and compared with immunohistochemical results. A complete scleral cartilage similar to that seen in birds, reptiles and amphibians encloses the platypus eye. This feature is present in sharks and rays, and in extant relatives of tetrapods, the lungfishes. The choroid lacks a tapetum. The retina is largely avascular and is rod-dominated, with a minority of red- and blue- cone immunoreactive photoreceptors. Like marsupials and many nonmammalian vertebrates, cones contain clear inner segment droplets. Double cones were present, a feature not found in eutherian mammals or marsupials. Evaluation of opsins indicates that red and blue immunoreactive cone opsins, but not rhodopsin, are present in the most basal of the extant species examined, the Pacific hagfish. Rhodopsin appears in the Australian and South American lungfishes, establishing emergence of this pigment in an extant relative of tetrapods. Unlike eyes of eutherian mammals, the platypus eye has retained morphologic features present in early tetrapods such as amphibians and their evolutionarily basal sister group, the lungfishes. These include scleral cartilage, double cones and cone droplets. In the platypus, as in other mammals, rod rhodopsin is the predominant photoreceptor pigment, at expense of the cone system.
Sigma-Aldrich
Anti-Rhodopsin Antibody, clone RET-P1, clone RET-P1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Opsin Antibody, blue, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Opsin Antibody, Red/Green, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Rhodopsin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution