Merck
  • Modulation of Microglia by Voluntary Exercise or CSF1R Inhibition Prevents Age-Related Loss of Functional Motor Units.
Modulation of Microglia by Voluntary Exercise or CSF1R Inhibition Prevents Age-Related Loss of Functional Motor Units.
Cell reports (2019-11-07)
Elisa Giorgetti, Moh Panesar, Yunyu Zhang, Stefanie Joller, Marie Ronco, Michael Obrecht, Christian Lambert, Nathalie Accart, Nicolau Beckmann, Arno Doelemeyer, Ludovic Perrot, Isabelle Fruh, Matthias Mueller, Eliane Pierrel, Serge Summermatter, Michael Bidinosti, Derya R Shimshek, Sophie Brachat, Mark Nash
ABSTRACT
Age-related loss of skeletal muscle innervation by motor neurons leads to impaired neuromuscular function and is a well-established clinical phenomenon. However, the underlying pathogenesis remains unclear. Studying mice, we find that the number of motor units (MUs) can be maintained by counteracting neurotoxic microglia in the aged spinal cord. We observe that marked innervation changes, detected by motor unit number estimation (MUNE), occur prior to loss of muscle function in aged mice. This coincides with gene expression changes indicative of neuronal remodeling and microglial activation in aged spinal cord. Voluntary exercise prevents loss of MUs and reverses microglia activation. Depleting microglia by CSF1R inhibition also prevents the age-related decline in MUNE and neuromuscular junction disruption, implying a causal link. Our results suggest that age-related changes in spinal cord microglia contribute to neuromuscular decline in aged mice and demonstrate that removal of aged neurotoxic microglia can prevent or reverse MU loss.
Sigma-Aldrich
Anti-Neurofilament 200kDa Antibody, clone NE14, Alexa Fluor 555 Conjugate, clone NE14, from mouse, ALEXA FLUOR 555
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Anti-NeuN Antibody (rabbit), from rabbit, purified by affinity chromatography
Roche
DAPI, 4′,6-Diamidine-2′-phenylindole dihydrochloride