Merck
  • Straight and branched-chain fatty acids in preorbital glands of sika deer, Cervus nippon.
Straight and branched-chain fatty acids in preorbital glands of sika deer, Cervus nippon.
Journal of chemical ecology (2004-04-29)
William F Wood
ABSTRACT
Using GC-MS analysis, 11 major volatile compounds were found in the preorbital gland secretion from a female sika deer, Cervus nippon. These compounds are the C14 through C18 straight-chain fatty acids, (ZZ)-9,12-octadecadienoic acid, 12-methyltridecanoic acid, 13-methyltetradecanoic acid, 14-methylpentadecanoic acid, 14-methylhexadecanoic acid, and 15-methylhexadecanoic acid. The five branched-chain acids make up over 29% of the volatiles in the gland. This is the first time branched-chain carboxylic acids have been reported from ungulate preorbital glands.
Sigma-Aldrich
12-Methyltridecanoic acid, 98% (capillary GC)
Sigma-Aldrich
Isopalmitic acid, ≥98% (capillary GC)